Assay Design



Gene Transcripts

Gene Symbol
beat-VII
Entrez Gene ID
43213
Gene Name
beaten path VII
Gene Aliases
Beat-VII, CG14248, CG14249, CG42242, Dmel\CG42242, Dmel_CG42242, beat VII
Chromosome Location
Chr.3R: 26645282 - 26681966 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0250908
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170283.4 NP_733162.2 4 - 5 930 112
GenBank mRNA BT029066.1 - 4 - 5 930 112

View Full Product Details