Assay Design
Gene Transcripts
- Gene Symbol
- beat-VII
- Entrez Gene ID
-
43213
- Gene Name
- beaten path VII
- Gene Aliases
-
Beat-VII, CG14248, CG14249, CG42242, Dmel\CG42242, Dmel_CG42242, beat VII
- Chromosome Location
-
Chr.3R: 26645282 - 26681966 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0250908
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170283.4 |
NP_733162.2 |
5 - 6 |
1078 |
58 |
GenBank mRNA |
BT029066.1 |
- |
5 - 6 |
1078 |
58 |
View Full Product Details