Assay Design



Gene Transcripts

Gene Symbol
dsd
Entrez Gene ID
43315
Gene Name
distracted
Gene Aliases
CG5634, CT17798, Dmel\CG5634, Dmel_CG5634
Chromosome Location
Chr.3R: 27597563 - 27607396 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039528
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276084.1 NP_001263013.1 5 - 6 3998 85
NM_143314.4 NP_651571.3 5 - 6 3998 85

View Full Product Details