Assay Design
Gene Transcripts
- Gene Symbol
- Gfat2
- Entrez Gene ID
-
43373
- Gene Name
- Glutamine:fructose-6-phosphate aminotransferase 2
- Gene Aliases
-
CG1345, Dmel/GFAT2, Dmel\CG1345, Dmel_CG1345, GFAT2, gfat2
- Chromosome Location
-
Chr.3R: 28337019 - 28339765 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039580
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143360.4 |
NP_651617.1 |
1 - 2 |
1891 |
61 |
GenBank mRNA |
AY095175.1 |
- |
1 - 2 |
1861 |
61 |
View Full Product Details