Assay Design



Gene Transcripts

Gene Symbol
ppk30
Entrez Gene ID
43487
Gene Name
pickpocket 30
Gene Aliases
CG18110, Dmel\CG18110, Dmel_CG18110
Chromosome Location
Chr.3R: 29669149 - 29671478 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039677
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276147.1 NP_001263076.1 2 - 3 682 62
NM_143449.2 NP_651706.1 2 - 3 682 62

View Full Product Details