Assay Design
Gene Transcripts
- Gene Symbol
- ppk19
- Entrez Gene ID
-
43489
- Gene Name
- pickpocket 19
- Gene Aliases
-
CG18287, Dmel\CG18287, Dmel_CG18287, PPK19
- Chromosome Location
-
Chr.3R: 29672456 - 29674223 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039679
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143451.2 |
NP_651708.2 |
2 - 3 |
974 |
99 |
View Full Product Details