Assay Design
Gene Transcripts
- Gene Symbol
- Cap-D2
- Entrez Gene ID
-
43491
- Gene Name
- CAP-D2 condensin subunit
- Gene Aliases
-
CAP-D2, CG1911, CapD2, DmCAP-D2, Dmel\CG1911, Dmel_CG1911, dCap-D2
- Chromosome Location
-
Chr.3R: 29675651 - 29680257 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039680
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143452.3 |
NP_651709.1 |
1 - 2 |
208 |
109 |
GenBank mRNA |
AY122183.1 |
- |
1 - 2 |
183 |
109 |
View Full Product Details