Assay Design
Gene Transcripts
- Gene Symbol
- PH4alphaSG2
- Entrez Gene ID
-
43623
- Gene Name
- prolyl-4-hydroxylase-alpha SG2
- Gene Aliases
-
CG1546, Dmel\CG1546, Dmel_CG1546, SG2
- Chromosome Location
-
Chr.3R: 30490074 - 30491987 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039779
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143544.3 |
NP_651801.1 |
3 - 4 |
1382 |
108 |
GenBank mRNA |
AF495537.1 |
- |
3 - 4 |
1343 |
108 |
View Full Product Details