Assay Design
Gene Transcripts
- Gene Symbol
- Rpt6R
- Entrez Gene ID
-
43635
- Gene Name
- Regulatory particle triple-A ATPase 6-related
- Gene Aliases
-
CG2241, DmCG2241, Dm_Rpt6b, Dmel\CG2241, Dmel_CG2241, Rpt6, Rpt6b, S8
- Chromosome Location
-
Chr.3R: 30582198 - 30583697 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039788
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143554.2 |
NP_651811.1 |
1 - 2 |
323 |
97 |
GenBank mRNA |
AY119229.1 |
- |
1 - 2 |
287 |
97 |
View Full Product Details