Assay Design



Gene Transcripts

Gene Symbol
VhaPPA1-1
Entrez Gene ID
45247
Gene Name
Vacuolar H[+] ATPase PPA1 subunit 1
Gene Aliases
CG7007, Dmel\CG7007, Dmel_CG7007, V0c'', Vha26, VhaPPA1, vhaPPA1-1
Chromosome Location
Chr.3R: 14902330 - 14904061 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028662
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260182.2 NP_001247111.1 2 - 3 788 86
NM_143753.2 NP_652010.1 2 - 3 792 86
GenBank mRNA AY060235.1 - 2 - 3 740 86

View Full Product Details