Assay Design



Gene Transcripts

Gene Symbol
Invadolysin
Entrez Gene ID
49580
Gene Name
CG3953-PA
Gene Aliases
CG3953, DmIX-14, Dmel\CG3953, Dmel_CG3953, IX-14, inv, invadolysin, l(3)IX-14, l(3)SG34
Chromosome Location
Chr.3R: 10144668 - 10159134 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086359
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143815.4 NP_652072.1 3 - 4 1046 84
GenBank mRNA AF234630.1 - 3 - 4 313 84
BT001861.1 - 3 - 4 1015 84

View Full Product Details