Assay Design



Gene Transcripts

Gene Symbol
Nmdar1
Entrez Gene ID
40665
Gene Name
NMDA receptor 1
Gene Aliases
CG2902, DMNMDAR1, DNMDAR-I, DNmdar, DmelNmdar1, Dmel\CG2902, Dmel_CG2902, DrNR1, NMDA, NMDA R1, NMDA-R1, NMDAR, NMDAR1, NMdar1, NR1, NRI, NmdaR1, Nmdar, dNR1, dnmdar-I, dsNR1, nmdar1, nmr, nr1
Chromosome Location
Chr.3R: 5508842 - 5523328 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010399
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_169059.2 NP_730940.1 10 - 11 1817 80
GenBank mRNA AY070577.1 - 10 - 11 1813 80
X71790.1 - - 1725 80

View Full Product Details