Assay Design
Gene Transcripts
- Gene Symbol
- Nmdar1
- Entrez Gene ID
-
40665
- Gene Name
- NMDA receptor 1
- Gene Aliases
-
CG2902, DMNMDAR1, DNMDAR-I, DNmdar, DmelNmdar1, Dmel\CG2902, Dmel_CG2902, DrNR1, NMDA, NMDA R1, NMDA-R1, NMDAR, NMDAR1, NMdar1, NR1, NRI, NmdaR1, Nmdar, dNR1, dnmdar-I, dsNR1, nmdar1, nmr, nr1
- Chromosome Location
-
Chr.3R: 5508842 - 5523328 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010399
View Full Product Details