Assay Design
Gene Transcripts
- Gene Symbol
- Jupiter
- Entrez Gene ID
-
41392
- Gene Name
- CG31363-PA
- Gene Aliases
-
BEST:LP05168, BcDNA:GH06373, BcDNA:LD21358, BcDNA:LD43690, BcDNA:RH53211, CG17238, CG18762, CG18763, CG31363, Dmel\CG31363, Dmel_CG31363, anon-EST:Liang-1.76, anon-WO0140519.173, clone 1.76, jupiter
- Chromosome Location
-
Chr.3R: 11590386 - 11619906 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0051363
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169409.4 |
NP_731600.1 |
2 - 3 |
657 |
60 |
GenBank mRNA |
AY069089.1 |
- |
2 - 3 |
640 |
60 |
View Full Product Details