Assay Design
Gene Transcripts
- Gene Symbol
- Osi22
- Entrez Gene ID
-
41632
- Gene Name
- Osiris 22
- Gene Aliases
-
CG8644, Dmel\CG8644, Dmel_CG8644
- Chromosome Location
-
Chr.3R: 13290629 - 13291846 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038133
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169501.2 |
NP_731783.1 |
1 - 2 |
764 |
91 |
View Full Product Details