Assay Design



Gene Transcripts

Gene Symbol
Ubx
Entrez Gene ID
42034
Gene Name
Ultrabithorax
Gene Aliases
BX-C, Bxl, CG10388, Cbx, DUbx, Dm Ubx, DmUbx, Dmel\CG10388, Dmel_CG10388, Hm, UBX, UBx, Uab, Ubx1b, abx, bx, bx[D], bxd, bxd[D], bxl, l(3)89Eb, pbx, ubx
Chromosome Location
Chr.3R: 16656623 - 16734426 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003944
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080504.4 NP_536752.1 3 - 4 1828 59
NM_169728.3 NP_732171.1 2 - 3 1750 59
NM_169729.3 NP_732172.1 3 - 4 1801 59
NM_169730.3 NP_732173.1 2 - 3 1777 59
GenBank mRNA BT010241.1 - 3 - 4 1801 59
BT125062.1 - 2 - 3 1729 59
X76210.1 - - 2028 59

View Full Product Details