Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038972.4 | NP_001034061.2 | 4 - 5 | 896 | 87 |
NM_001170184.2 | NP_001163655.1 | 4 - 5 | 896 | 87 | |
NM_001300479.1 | NP_001287408.1 | 4 - 5 | 896 | 87 | |
NM_001300480.1 | NP_001287409.1 | 4 - 5 | 896 | 87 | |
NM_001300481.1 | NP_001287410.1 | 4 - 5 | 896 | 87 | |
NM_169873.4 | NP_732447.2 | 4 - 5 | 896 | 87 |