Assay Design



Gene Transcripts

Gene Symbol
GluRIIE
Entrez Gene ID
318623
Gene Name
Glutamate receptor IIE
Gene Aliases
CG31201, DmelGluRIIE, Dmel\CG31201, Dmel_CG31201, GluR-IIE
Chromosome Location
Chr.3R: 20644154 - 20648716 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051201
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043268.2 NP_001036733.1 1 - 2 244 79
GenBank mRNA BT056325.1 - 1 - 2 242 79

View Full Product Details