Assay Design
Gene Transcripts
- Gene Symbol
- Gr93d
- Entrez Gene ID
-
117470
- Gene Name
- Gustatory receptor 93d
- Gene Aliases
-
93F.4, CG31335, DmGr93d, Dmel\CG31335, Dmel_CG31335, GR93F.4, GR93d, Gr93F4
- Chromosome Location
-
Chr.3R: 21845454 - 21846659 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0045468
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169974.1 |
NP_732666.1 |
1 - 2 |
983 |
72 |
View Full Product Details