Assay Design



Gene Transcripts

Gene Symbol
fzo
Entrez Gene ID
42745
Gene Name
fuzzy onions
Gene Aliases
CG4568, Dmel\CG4568, Dmel_CG4568, Fzo, MFN1, anon-WO0125274.1, anon-WO0125274.25, anon-WO0125274.26, mfn1
Chromosome Location
Chr.3R: 23201644 - 23204034 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011596
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170060.2 NP_732840.1 1 - 1 2159 107
GenBank mRNA BT023200.1 - 1 - 1 2156 107
BT023220.1 - 1 - 1 2158 107
BT023232.1 - 1 - 1 2156 107
U95821.1 - 1 - 1 2159 107

View Full Product Details