Assay Design
Gene Transcripts
- Gene Symbol
- Lsd-1
- Entrez Gene ID
-
42810
- Gene Name
- Lipid storage droplet-1
- Gene Aliases
-
CG10374, DmPLIN1, Dmel\CG10374, Dmel_CG10374, LSD-1, LSD1, LSDP1, Lsd1, Lsdp1, PLIN1, Plin1, dmPLIN1, lsd-1, lsd1, plin1
- Chromosome Location
-
Chr.3R: 23763857 - 23766662 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039114
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170092.3 |
NP_732904.2 |
4 - 5 |
668 |
73 |
View Full Product Details