Assay Design



Gene Transcripts

Gene Symbol
Rox8
Entrez Gene ID
42848
Gene Name
CG5422-PB
Gene Aliases
CG5422, DmTIAR, Dmel\CG5422, Dmel_CG5422, ROX8, RRM12, Rbp12, Rox-8, TIAR, dTIAR, rox8, rrm12
Chromosome Location
Chr.3R: 24043056 - 24048265 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005649
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275968.1 NP_001262897.1 3 - 4 2143 55
NM_001275969.1 NP_001262898.1 3 - 4 2185 55
NM_170116.2 NP_732942.1 3 - 4 2051 55
NM_170117.2 NP_732943.1 3 - 4 2131 55
NM_170118.2 NP_732944.2 3 - 4 2037 55
NM_170119.2 NP_732945.1 3 - 4 2115 55
GenBank mRNA BT050464.1 - 3 - 4 2115 55

View Full Product Details