Assay Design



Gene Transcripts

Gene Symbol
nAChRbeta2
Entrez Gene ID
42920
Gene Name
nicotinic Acetylcholine Receptor beta2
Gene Aliases
Acr96Ac, AcrF, CG6798, Dbeta2, Dmel\CG6798, Dmel_CG6798, EMS2, SBD, beta2, beta[[2]], nACR beta 96A, nACRbeta, nAChR, nAChR-beta96A, nAcRbeta, nAcRbeta-96A, nAcRs, nAcr96Ac, sbd
Chromosome Location
Chr.3R: 24509228 - 24515106 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004118
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079759.4 NP_524483.1 4 - 5 704 64
NM_170147.3 NP_733002.1 4 - 5 704 64
GenBank mRNA BT133158.1 - 4 - 5 684 64

View Full Product Details