Assay Design



Gene Transcripts

Gene Symbol
slo
Entrez Gene ID
42940
Gene Name
slowpoke
Gene Aliases
BcDNA:GH10751, CG10693, Dmel\CG10693, Dmel_CG10693, Dslo, Slo, Slo1, dSlo, dSlo1, dslo, fSlo
Chromosome Location
Chr.3R: 24662491 - 24711595 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003429
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014654.3 NP_001014654.1 15 - 16 2699 69
NM_170164.4 NP_733029.1 16 - 17 2440 69
NM_206566.4 NP_996289.1 16 - 17 2440 69

View Full Product Details