Assay Design
Gene Transcripts
- Gene Symbol
- RabX4
- Entrez Gene ID
-
42960
- Gene Name
- CG31118-PA
- Gene Aliases
-
AAF56345, CG13638, CG31118, DmRabX4, Dmel\CG31118, Dmel_CG31118, rabX4
- Chromosome Location
-
Chr.3R: 24826665 - 24828409 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0051118
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170170.3 |
NP_733043.1 |
2 - 3 |
873 |
83 |
View Full Product Details