Assay Design
Gene Transcripts
- Gene Symbol
- vig2
- Entrez Gene ID
-
43016
- Gene Name
- CG11844-PA
- Gene Aliases
-
BcDNA:SD06613, CG11844, Dmel\CG11844, Dmel_CG11844, Vig2, anon-WO0118547.290, vig-2
- Chromosome Location
-
Chr.3R: 25123373 - 25125619 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0046214
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170204.2 |
NP_733083.1 |
1 - 2 |
110 |
77 |
View Full Product Details