Assay Design
Gene Transcripts
- Gene Symbol
- ymp
- Entrez Gene ID
-
43088
- Gene Name
- yellow-emperor
- Gene Aliases
-
BcDNA:AT03213, CG12250, Dmel\CG12250, Dmel_CG12250
- Chromosome Location
-
Chr.3R: 25591502 - 25609127 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261287
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170230.2 |
NP_733109.1 |
5 - 6 |
511 |
81 |
View Full Product Details