Assay Design
Gene Transcripts
- Gene Symbol
- fd96Cb
- Entrez Gene ID
-
43011
- Gene Name
- forkhead domain 96Cb
- Gene Aliases
-
CG11922, Dm-FoxB2, Dmel\CG11922, Dmel_CG11922, Dmfd5, FD5, fd5
- Chromosome Location
-
Chr.3R: 25095012 - 25096099 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004898
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079772.2 |
NP_524496.1 |
1 - 1 |
814 |
116 |
GenBank mRNA |
BT028866.2 |
- |
1 - 1 |
708 |
116 |
View Full Product Details