Assay Design
Gene Transcripts
- Gene Symbol
- stg
- Entrez Gene ID
-
43466
- Gene Name
- string
- Gene Aliases
-
0224/06, 0245/03, 0439/22, 0730/13, 0896/05, 0967/05, 0980/06, 1083/13, 1089/08, 1143/02, 5473, CDC25, CDC25[string], CG1395, Cdc25, Cdc25[Stg], Cdc25[String], Cdc25[stg], Cdc25[string], Cdc25c, Dmel\CG1395, Dmel_CG1395, EP1213, S(rux)3A, STG, SY3-4, Stg, Stg/Cdc25, String/Cdc25, anon-EST:Liang-2.21, cdc25, cdc25[string], clone 2.21, l(3)01235, l(3)j10B9, l(3)j1D3, l(3)j1E3, l(3)j3D1, l(3)s2213, st, stg[cdc25], str/cdc25
- Chromosome Location
-
Chr.3R: 29251826 - 29255800 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003525
View Full Product Details