Assay Design



Gene Transcripts

Gene Symbol
TkR99D
Entrez Gene ID
43551
Gene Name
Tachykinin-like receptor at 99D
Gene Aliases
CG7887, DTKR, DmTRPR, Dmel\CG7887, Dmel_CG7887, Dtkr, TAKR99D, TKR99D, TLR2_DROME, Tak-r, TakR99D, Takr99D, anon-WO0131005.11, anon-WO0131005.9, dtkr, l(3)s2222
Chromosome Location
Chr.3R: 29950796 - 29969447 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004622
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170301.1 NP_001163772.1 3 - 4 1032 55
NM_001276163.1 NP_001263092.1 3 - 4 1032 55
NM_079832.3 NP_524556.2 3 - 4 1032 55
GenBank mRNA BT150106.1 - 3 - 4 1032 55

View Full Product Details