Assay Design



Gene Transcripts

Gene Symbol
CanA1
Entrez Gene ID
43670
Gene Name
Calcineurin A1
Gene Aliases
CAN A1/PP2B, CG1455, CNA1, CaN, Can, Cana1, CnA, CnnA21EF, D14, DmCanA1-100B, Dmel\CG1455, Dmel_CG1455, PP2B, PP2B 21EF, PpD14, canA, canA1, pMY4
Chromosome Location
Chr.3R: 31039374 - 31046821 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010015
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260449.2 NP_001247378.2 8 - 9 2048 59
NM_079861.4 NP_524600.3 9 - 10 1612 59
GenBank mRNA AY128480.1 - 9 - 10 1610 59

View Full Product Details