Assay Design



Gene Transcripts

Gene Symbol
chp
Entrez Gene ID
43690
Gene Name
chaoptin
Gene Aliases
24B10, 516, BSH, CG1744, CHP, CT5040, Chaoptin, Chp, Dmel\CG1744, Dmel_CG1744, MAb 24B10, MAb24B10, Mab24B10, Nsp, idl-A, mAB24B10, mAb 24B10, mAb24B10
Chromosome Location
Chr.3R: 31204180 - 31210730 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267435
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276205.1 NP_001263134.1 2 - 3 681 62
NM_001276206.1 NP_001263135.1 2 - 3 702 62
NM_001276207.1 NP_001263136.1 2 - 3 681 62
NM_001276208.1 NP_001263137.1 2 - 3 702 62
NM_079866.3 NP_524605.1 2 - 3 702 62
GenBank mRNA BT126010.1 - 2 - 3 480 62

View Full Product Details