Assay Design
Gene Transcripts
- Gene Symbol
- boss
- Entrez Gene ID
-
43146
- Gene Name
- bride of sevenless
- Gene Aliases
-
BOSS, Bos, Boss, CG8285, Dmel\CG8285, Dmel_CG8285, F, X55887
- Chromosome Location
-
Chr.3R: 25961994 - 25966948 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000206
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080709.3 |
NP_542440.1 |
2 - 3 |
223 |
63 |
GenBank mRNA |
BT088800.1 |
- |
2 - 3 |
208 |
63 |
View Full Product Details