Assay Design



Gene Transcripts

Gene Symbol
spn-D
Entrez Gene ID
318579
Gene Name
spindle D
Gene Aliases
BcDNA:RE19845, CG31069, Dmel\CG31069, Dmel_CG31069, Spn-D, rad51C, spnD, spndD
Chromosome Location
Chr.3R: 27165716 - 27167122 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003482
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170321.3 NP_733200.1 3 - 4 436 76
GenBank mRNA AY071162.1 - 3 - 4 434 76
AY257540.1 - 3 - 4 432 76

View Full Product Details