Assay Design



Gene Transcripts

Gene Symbol
PH4alphaPV
Entrez Gene ID
43640
Gene Name
prolyl-4-hydroxylase-alpha PV
Gene Aliases
CG31015, CG9713, Dmel\CG31015, Dmel_CG31015
Chromosome Location
Chr.3R: 30646106 - 30648551 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051015
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170516.1 NP_733395.1 5 - 6 1351 67
GenBank mRNA AF495539.1 - 5 - 6 1351 67
BT150220.1 - 5 - 6 1351 67

View Full Product Details