Assay Design



Gene Transcripts

Gene Symbol
dpr11
Entrez Gene ID
40800
Gene Name
defective proboscis extension response 11
Gene Aliases
BcDNA:GH22307, CG15181, CG15182, CG15183, CG31309, CG33202, CT35098, Dmel\CG33202, Dmel_CG33202, Dpr-11, Dpr11
Chromosome Location
Chr.3R: 6529169 - 6593330 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053202
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275391.1 NP_001262320.1 5 - 6 1722 56
NM_176409.2 NP_788586.1 5 - 6 1722 56
GenBank mRNA AY121627.1 - 5 - 6 1722 56

View Full Product Details