Assay Design
Gene Transcripts
- Gene Symbol
- Ppn
- Entrez Gene ID
-
43872
- Gene Name
- Papilin
- Gene Aliases
-
CG1540, CG18436, CG33103, CT3186, CT41988, Dmel\CG33103, Dmel_CG33103, ppl, ppn
- Chromosome Location
-
Chr.3R: 28520665 - 28540324 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003137
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176574.3 |
NP_788751.2 |
10 - 11 |
7137 |
59 |
View Full Product Details