Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170289.2 | NP_001163760.1 | 2 - 3 | 472 | 72 |
NM_001170290.2 | NP_001163761.1 | 2 - 3 | 472 | 72 | |
NM_176574.3 | NP_788751.2 | 2 - 3 | 472 | 72 | |
NM_176575.3 | NP_788752.2 | 2 - 3 | 472 | 72 |