Assay Design



Gene Transcripts

Gene Symbol
Ppn
Entrez Gene ID
43872
Gene Name
Papilin
Gene Aliases
CG1540, CG18436, CG33103, CT3186, CT41988, Dmel\CG33103, Dmel_CG33103, ppl, ppn
Chromosome Location
Chr.3R: 28520665 - 28540324 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003137
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170290.2 NP_001163761.1 8 - 9 6783 107
NM_176574.3 NP_788751.2 9 - 10 6954 107
NM_176575.3 NP_788752.2 9 - 10 6954 107

View Full Product Details