Assay Design



Gene Transcripts

Gene Symbol
Doa
Entrez Gene ID
43415
Gene Name
Darkener of apricot
Gene Aliases
0844/02, CG1658, CG31049, CG33204, CG33553, CG42320, DOA, DOA/CLK2, Dmel\CG42320, Dmel_CG42320, Msu, doa, l(3)01705, l(3)S084402, l(3)s2784
Chromosome Location
Chr.3R: 28888073 - 28922772 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265998
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260425.1 NP_001247354.1 2 - 3 757 106
NM_176581.4 NP_788758.3 2 - 3 757 106

View Full Product Details