Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170223.1 | NP_001163694.1 | 5 - 6 | 811 | 69 |
NM_206544.2 | NP_996267.1 | 5 - 6 | 586 | 69 | |
NM_206545.2 | NP_996268.1 | 4 - 5 | 293 | 69 | |
GenBank mRNA | AF192555.1 | - | 4 - 5 | 277 | 69 |
AF224713.1 | - | 4 - 5 | 270 | 69 | |
AF224714.1 | - | 4 - 5 | 277 | 69 | |
AF244918.1 | - | 4 - 5 | 277 | 69 | |
AF250918.1 | - | 4 - 5 | 277 | 69 | |
AF263722.1 | - | 4 - 5 | 293 | 69 | |
AY069096.1 | - | 4 - 5 | 277 | 69 | |
DQ191318.1 | - | 5 - 6 | 526 | 69 |