Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032033.2 | NP_001027204.2 | 10 - 11 | 2360 | 99 |
NM_001032035.2 | NP_001027206.1 | 9 - 10 | 2479 | 99 | |
NM_001032036.2 | NP_001027207.1 | 10 - 11 | 2376 | 99 | |
NM_001032037.2 | NP_001027208.1 | 10 - 11 | 2493 | 99 | |
NM_206555.2 | NP_996278.1 | 9 - 10 | 2463 | 99 | |
NM_206556.2 | NP_996279.1 | 9 - 10 | 2463 | 99 | |
GenBank mRNA | AF454350.1 | - | 9 - 10 | 2463 | 99 |
AF454351.1 | - | 9 - 10 | 2463 | 99 | |
BT011452.1 | - | 10 - 11 | 2373 | 97 |