Assay Design
Gene Transcripts
- Gene Symbol
- Hsc70-2
- Entrez Gene ID
-
41609
- Gene Name
- Heat shock protein cognate 2
- Gene Aliases
-
CG7756, Dmel\CG7756, Dmel_CG7756, HSC-70, HSC2, HSC70, HSC70-2, HSP7B_DROME, Hsc-2, Hsc2, Hsc70, Hsc70.2, dhsc70, hsc2, hsc70
- Chromosome Location
-
Chr.3R: 13044759 - 13047388 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001217
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079615.4 |
NP_524339.1 |
1 - 2 |
368 |
130 |
View Full Product Details