Assay Design
Gene Transcripts
- Gene Symbol
- Obp56g
- Entrez Gene ID
-
37270
- Gene Name
- Odorant-binding protein 56g
- Gene Aliases
-
56g, CG13873, DmelObp56g, Dmel\CG13873, Dmel_CG13873, OBP56g
- Chromosome Location
-
Chr.2R: 19783484 - 19784043 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034474
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137603.3 |
NP_611447.1 |
1 - 2 |
85 |
137 |
View Full Product Details