Assay Design
Gene Transcripts
- Gene Symbol
- Acp26Aa
- Entrez Gene ID
-
33817
- Gene Name
- Accessory gland protein 26Aa
- Gene Aliases
-
Acp, Acp26A, Acp26Aab, Acp26a, CG8982, Dmel\CG8982, Dmel_CG8982, Mst26A, Mst26Aa, Ovulin, msP355a, mst 355a, mst355a, ovulin
- Chromosome Location
-
Chr.2L: 5892883 - 5893896 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002855
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057296.2 |
NP_476644.1 |
2 - 2 |
898 |
150 |
GenBank mRNA |
BT024423.1 |
- |
2 - 2 |
873 |
150 |
View Full Product Details