Assay Design
Gene Transcripts
- Gene Symbol
- cpb
- Entrez Gene ID
-
33346
- Gene Name
- capping protein beta
- Gene Aliases
-
CAPB_DROME, CG17158, CP, CP-beta, CPB, Cap2, CapZ, CapZbeta, Cpb, Dmel\CG17158, Dmel_CG17158, J, bnd, cpbeta
- Chromosome Location
-
Chr.2L: 1704902 - 1706962 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011570
View Full Product Details