Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001274761.1 | NP_001261690.1 | 3 - 3 | 3829 | 136 |
NM_001274762.1 | NP_001261691.1 | 3 - 3 | 3829 | 136 | |
NM_001300095.1 | NP_001287024.1 | 3 - 3 | 4184 | 136 | |
NM_057714.4 | NP_477062.1 | 3 - 3 | 3829 | 136 | |
GenBank mRNA | BT025225.1 | - | 3 - 3 | 3675 | 136 |