Assay Design
Gene Transcripts
- Gene Symbol
- DnaJ-1
- Entrez Gene ID
-
38643
- Gene Name
- DnaJ-like-1
- Gene Aliases
-
CG10578, DNAJ-1, DNAJ1, DROJ1, DmHsp40, Dmel\CG10578, Dmel_CG10578, DnaJ1, DnaJ1 64EF, Dnaj-1, DroJ1, EU3500, HSP40, HSP40/HDJ1, Hdj1, Hsp-40, Hsp40, anon-WO0140519.166, anon-WO0172774.135, dHDJ1, dHDJ1/HSP40, dHSP40, dHdj1, dhdJ1, dhdj-1, dhdj1, dnaJ-1, dnaj-1, droj1, hsp40
- Chromosome Location
-
Chr.3L: 5750029 - 5752189 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0263106
View Full Product Details