Assay Design
Gene Transcripts
- Gene Symbol
- LSm7
- Entrez Gene ID
-
35003
- Gene Name
- Like Sm 7
- Gene Aliases
-
BcDNA:RH73529, CG13277, Dmel\CG13277, Dmel_CG13277, LSM7, Lsm7
- Chromosome Location
-
Chr.2L: 16742315 - 16743100 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261068
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135963.4 |
NP_609807.1 |
2 - 2 |
457 |
120 |
GenBank mRNA |
AY113637.1 |
- |
2 - 2 |
447 |
120 |
View Full Product Details