Assay Design
Gene Transcripts
- Gene Symbol
- bc10
- Entrez Gene ID
-
38628
- Gene Name
- CG4867-PA
- Gene Aliases
-
CG4867, Dmel\CG4867, Dmel_CG4867, dbc10
- Chromosome Location
-
Chr.3L: 5565231 - 5566765 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040239
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139715.6 |
NP_647972.1 |
2 - 2 |
1251 |
108 |
GenBank mRNA |
AY128448.1 |
- |
2 - 2 |
1219 |
108 |
View Full Product Details